missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Reelin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5149714
This item is not returnable.
View return policy
Description
Immunogen sequence: QYSNDNGILW HLLRELDFMS FLEPQIISID LPQDAKTPAT AFRWWQPQHG KHSAQWALDD VLIGMNDSSQ TGFQDKFDGS IDLQANWYRI QGGQVDIDCL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.
Specifications
| Reelin | |
| Polyclonal | |
| Unconjugated | |
| RELN | |
| ETL7; extracellular matrix serine protease; LIS2; PRO1598; Reelen; reeler; Reeler protein; Reelin; RELN; Rl | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5649 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78509 | |
| RELN | |
| Recombinant protein corresponding to Human RELN. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction