Learn More
Thermo Scientific™ Solid tumor: AcroMetrix Oncology Hotspot Control (IVD, CE-IVD)
Product Code. 15902951 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 x 25 μL
Unit Size:
75µL
Product Code. Quantity Unit Size
15902951 3 x 25 μL 75µL
1 options
Product Code. 15902951 Supplier Thermo Scientific™ Supplier No. 969056
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Assess day-to-day test variation of somatic mutation panels with the Thermo Scientific™ AcroMetrix™ Oncology Hotspot Control, which is for In Vitro Diagnostic Use.
Assess day-to-day test variation of somatic mutation panels. Intended for use with next generation sequencing (NGS) assays designed to identify somatic mutations in DNA from human samples. For In Vitro Diagnostic Use.
  • Multiple variant types to capture the diversity of somatic cancer mutation types
  • 500+ clinically relevant variants allow detection of multiple targets with NGS in a single reaction
  • IVD labeled enables use in clinical diagnostics
  • Variant frequency near LOD for test method development and validation
FAQs
TRUSTED_SUSTAINABILITY
Specifications
Type Oncology Hotspot Control
Packaging 3 x 25µL
Quantity 3 x 25 μL
compliance-icons
Product Identifier
  • AcroMetrix Oncology Hotspot Control
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tris{hydroxyméthyl}aminométhane