missing translation for 'onlineSavingsMsg'
Learn More

ONC212, MedChemExpress

Product Code. 16437147
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437147

Brand: Medchem Express HY111343

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ONC212, a fluorinated-ONC201 analogue, is a promising anti-cancer agent and also a selective agonist of GPR132. ONC212 also induces apoptosis.

ONC212, a fluorinated-ONC201 analogue, is a promising anti-cancer agent and also a selective agonist of GPR132. ONC212 also induces apoptosis.

Specifications

Chemical Name or Material ONC212
Physical Form Solid
CAS 1807861-48-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H23F3N4O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (113.52 mM; Need ultrasonic)
SMILES O=C1N(CC2=CC=C(C(F)(F)F)C=C2)C3=NCCN3C4=C1CN(CC5=CC=CC=C5)CC4
Molecular Weight (g/mol) 440.46
Formula Weight 440.46
Percent Purity 99.84%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.