missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Carlo Erba On-Column Syringe

On-column syringes require the needle OD to be smaller than the ID of the capillary column with which it is used. For use with Carlo Erba products

£68.00 - £109.00

Specifications

Diameter (Metric) Outer 0.23 mm
Product Type On Column Syringe
Length (Metric) Needle 75 mm
View More Specs

Products 2
Product Code Brand Volume (Metric) Length (Metric) Needle Price Quantity & Availability  
Product Code Brand Volume (Metric) Length (Metric) Needle Price Quantity & Availability  
10626231
View accessory products View Documents Promotion Details
Trajan™
000380
5 μL 75 mm
£109.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10015510
View accessory products View Documents Promotion Details
Trajan™
002500
10 μL 75 mm
£68.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Designed for on-column injections
Specifications

Specifications

0.23 mm
On Column Syringe
75 mm
Videos
SDS
Documents

Documents

Product Certifications