missing translation for 'onlineSavingsMsg'
Learn More

Hexamethonium Bromide, MedChemExpress

Product Code. 16411008
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16411008

Brand: Medchem Express HYB0569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) antagonist, with mixed competitive and noncompetitive activity. Hexamethonium Bromide has anti-hypertensive activity. Hexamethonium Bromide attenuates sympathetic activity and blood pressure in spontaneously hypertensive animal models.

Hexamethonium Bromide is a non-selective ganglionic nicotinic-receptor antagonist (nAChR) antagonist, with mixed competitive and noncompetitive activity. Hexamethonium Bromide has anti-hypertensive activity. Hexamethonium Bromide attenuates sympathetic activity and blood pressure in spontaneously hypertensive animal models.

Specifications

Chemical Name or Material Hexamethonium Bromide
Physical Form Solid
CAS 55-97-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C12H30Br2N2
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information H2O : ≥ 100 mg/mL (276.10 mM) ∣DMSO : 16.67 mg/mL (46.03 mM; Need ultrasonic)
SMILES C[N+](C)(C)CCCCCC[N+](C)(C)C.[Br-].[Br-]
Molecular Weight (g/mol) 362.19
Formula Weight 362.19
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.