missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diba Omnifit™ Tube End Fittings
Click to view available options
Connections:
Tube End Fit 1/4-28 F/1/16 in.
Tube End Fit 1/4-28 F/1/8 in.
Unit Size:
Pack of 10
Description
- Won't compress tubing; allow it to be free spinning
- Must be used with Gripper Fittings
Specifications
Specifications
| Type | Tube End Fitting |
| Connections | Tube End Fit 1/4-28 F/1/8 in. |
| Material | Polypropylene |
| Color | Assorted |
| For Use With (Equipment) | 1/8 in. O.D. Tubing |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction