missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ T-Series Bottle Caps

Product Code. 15224579
Encompass_Preferred
Click to view available options
Thread Size (Metric):
GL 32
GL 38
GL 45
Includes:
2 Ports
2 Ports, Check Valve
2 Ports, No Valve
3 Ports
3 Ports, Check Valve
3 Ports, No Valve
4 Ports, Check Valve
4 Ports, No Valve
Four port
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15224579

Brand: Diba 00938T2F

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Easy to use and versatile range of caps requires no fittings

  • Luer ported, straight through tubing
  • Simply push 1/8 in. O.D. hard wall tubing through the ports
  • For 1/16 in. O.D. hard wall tubing, use OF9LA adapter supplied with cap
  • Use OF9BA series adapter for softwall tubing
  • Anti-twist design prevents tubing from kinking when removing cap
  • Each port also accepts male Luer connectors
  • With polypropylene screw cap and chemically inert solid PTFE body

Not recommended for use under pressure with Luer products fitting as no Luer lock is employed.

Order Info

Available with Luer port or Luer port with fluorocarbon check valve and 10μm porosity PTFE inlet filter enabling air to flow into the bottle while preventing particulate contamination and preventing harmful vapors from escaping.

Specifications

Color Blue, White
Includes 2 Ports, Check Valve
Material PTFE
No. per Case 1 Cs.
Sterility Non-sterile
Thread Size (Metric) GL 38
Neck Finish 38-430
Disposable Reusable
Height (Metric) 28.5 mm
Product Type Solvent Waste Cap
For Use With (Equipment) T series solvent bottle
Diameter (Metric) 54 mm
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.