Learn More
Diba Omnifit™ Packing Sleeves
Product Code. 15254509
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
10 mm (Bore Size) Columns
15 mm (Bore Size) Columns
25 mm (Bore Size) Columns
3.0 mm (Bore Size) Columns
50 mm (Bore Size) Columns
6.6 mm (Bore Size) Columns
Unit Size:
Each
Product Code. 15254509
Brand: Diba 006PS03
This item requires a price update prior to being ordered.
Request a Price for this item here.
Sleeves to connect glass columns
  • For Omnifit glass low/medium pressure liquid chromatography columns
  • Offered in multiple I.D. (bore) sizes
Specifications
Type Sleeve
Diameter (Metric) Inner 3.0 mm (Bore Size) Columns