Learn More
Diba Omnifit™ Cap Adapters
Product Code. p-3112341
Encompass_Preferred
Click to view available options
Connections:
Cap to Female Luer Adapter
Unit Size:
Each
Product Code. 15254439
Brand: Diba 001202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Adapters replace the cap on any Omnifit cap connector or valve
  • Enable quick push-on connections for softwall tubing or Luer connectors
  • Autoclavable
Specifications
Type Cap Adapter
Connections Cap to Female Luer Adapter
For Use With (Equipment) Female Luer Adapter