Learn More
Diba Omnifit™ Cap Adapters
Click to view available options
Connections:
Cap to Female Luer Adapter
Unit Size:
Each
Description
- Enable quick push-on connections for softwall tubing or Luer connectors
- Autoclavable
Specifications
Specifications
| Type | Cap Adapter |
| Connections | Cap to Female Luer Adapter |
| For Use With (Equipment) | Female Luer Adapter |