missing translation for 'onlineSavingsMsg'
Learn More

Specac™ Omni-Cell™ ZnSe Mull Windows Circular 25mm Diameter

Product Code. 15289837
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment)
15289837 Omni-Cell GS01800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15289837 Supplier Specac™ Supplier No. GS01834
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
ZnSe circular mull cell windows for use in the Omni-Cell GS01800. 25mm diameter and 2mm thick.

Specifications

For Use With (Equipment) Omni-Cell GS01800
Description 2 mm thick
Diameter (Metric) 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.