Learn More
Specac™ Omni-Cell™ ZnSe Liquid Cell Windows Rectangular
Product Code. 15289827
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
Product Code. For Use With (Equipment)
15289827 Omni-Cell GS01800
1 options
Product Code. 15289827 Supplier Specac™ Supplier No. GS01814
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
ZnSe rectangular liquid cell windows for use in the Omni-Cell GS01800. 41mm x 23mm and 2mm thick.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Description 41 mm x 23 mm and 2 mm thick