Learn More
Specac™ Omni-Cell™ Spacers PTFE Rectangular 0.05mm
Product Code. 15269847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Pk.
Product Code. Quantity
15269847 5 Pk.
1 options
Product Code. 15269847 Supplier Specac™ Supplier No. GS01850
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Packet of 5 rectangular PTFE spacers with a thickness of 0.05mm for use in the Omni-Cell GS01800.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Quantity 5 Pk.