Learn More
Specac™ Omni-Cell™ Spacers PTFE Circular 0.10mm for Mull Cells
Product Code. 15229867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Pk.
Product Code. Quantity
15229867 5 Pk.
1 options
Product Code. 15229867 Supplier Specac™ Supplier No. GS01871
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Packet of 5 circular PTFE spacers with a thickness of 0.10mm for use in the Omni-Cell GS01800.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Quantity 5 Pk.