Learn More
Specac™ Omni-Cell™ Spacers Mylar Rectangular Assorted Sizes
Description
Packet of 10 assorted rectangular PTFE spacers. 2 x 0.05mm, 2 x 0.10mm, 2 x 0.20mm, 2 x 0.50mm and 2 x 1.00mm for use in the Omni-Cell GS01800.
Specifications
Specifications
| For Use With (Equipment) | Omni-Cell GS01800 |
| Quantity | 10 Pk. |