Learn More
Specac™ Omni-Cell™ Spacers Mylar Rectangular 0.025mm
Product Code. 15299857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Pk.
Product Code. Quantity
15299857 5 Pk.
1 options
Product Code. 15299857 Supplier Specac™ Supplier No. GS01863
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Packet of 5 rectangular Mylar spacers with a thickness of 0.025mm for use in the Omni-Cell GS01800.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Quantity 5 Pk.