Learn More
Specac™ Omni-Cell™ Spacers Mylar Circular 0.006mm
Product Code. 15229877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Pk.
Product Code. Quantity
15229877 5 Pk.
1 options
Product Code. 15229877 Supplier Specac™ Supplier No. GS01881
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Packet of 5 circular Mylar spacers with a thickness of 0.006mm for use in the Omni-Cell GS01800.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Quantity 5 Pk.