Learn More
Specac™ Omni-Cell™ Spacers Lead Rectangular 0.05mm
Product Code. 15229857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Pk.
Product Code. Quantity
15229857 5 Pk.
1 options
Product Code. 15229857 Supplier Specac™ Supplier No. GS01856
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Packet of 5 rectangular lead spacers with a thickness of 0.05mm for use in the Omni-Cell GS01800.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Quantity 5 Pk.