Learn More
Specac™ Omni-Cell™ NaCl Liquid Cell Windows Rectangular
Product Code. 15249827
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15249827 Omni-Cell GS01800 Each
1 options
Product Code. 15249827 Supplier Specac™ Supplier No. GS01810
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
NaCl rectangular Liquid Cell windows for use in the Omni-Cell GS01800. 41mm x 23mm and 4mm thick.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Description 41 mm x 23 mm and 4 mm thick.