Learn More
Specac™ Omni-Cell™ KRS-5 Liquid Cell Windows Rectangular
Description
KRS-5 rectangular liquid cell windows for use in the Omni-Cell GS01800. 41mm x 23mm and 4mm thick.
Specifications
Specifications
| For Use With (Equipment) | Omni-Cell GS01800 |
| Description | 41 mm x 23 mm and 4 mm thick |