missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Omni-Cell™ KBr Sealed Window Unit 0.50mm
Click to view available options
Type:
Omni-Cell KBr Sealed Window Unit 0.50mm
Unit Size:
Each
Description
Permanently sealed Omni-Cell GS01800 with KBr rectangular windows and 0.50mm thickness lead spacer. Cannot be disassembled. Ideal for volatile liquids, quantitative applications and low viscosity liquids.
Specifications
Specifications
| Type | Omni-Cell KBr Sealed Window Unit 0.50mm |
| Description | 0.50 mm Thickness Lead Spacer |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction