Learn More
Specac™ Omni-Cell™ Fused Silica Sealed Window Unit 1.00mm
Description
Permanently sealed Omni-Cell GS01800 with fused silica rectangular windows and 1.00mm thickness lead spacer. Cannot be disassembled. Ideal for volatile liquids, quantitative applications and low viscosity liquids.
Specifications
Specifications
| Type | Omni-Cell Fused Silica Sealed Window Unit |
| Description | 1.00 mm Thickness Lead Spacer |