Learn More
Specac™ Omni-Cell™ Fused Silica Sealed Window Unit 0.20mm
Product Code. 15249917
Encompass_Preferred
Click to view available options
Type:
Omni-Cell Fused Silica Sealed Window Unit
Product Code. 15249917
Brand: Specac™ GS01948
This item requires a price update prior to being ordered.
Request a Price for this item here.
Permanently sealed Omni-Cell GS01800 with fused silica rectangular windows and 0.20mm thickness lead spacer. Cannot be disassembled. Ideal for volatile liquids, quantitative applications and low viscosity liquids.
Specifications
Type Omni-Cell Fused Silica Sealed Window Unit
Description 0.20 mm Thickness Lead Spacer