Learn More
Specac™ Omni-Cell™ Fused Silica Sealed Window Unit 0.05mm
Product Code. 15269897
Encompass_Preferred
Change view
Click to view available options
Type:
Omni-Cell Fused Silica Sealed Window Unit
Product Code. Type
15269897 Omni-Cell Fused Silica Sealed Window Unit
1 options
Product Code. 15269897 Supplier Specac™ Supplier No. GS01928
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Permanently sealed Omni-Cell GS01800 with fused silica rectangular windows and 0.05mm thickness lead spacer. Cannot be disassembled. Ideal for volatile liquids, quantitative applications and low viscosity liquids.
Specifications
Type Omni-Cell Fused Silica Sealed Window Unit
Description 0.05 mm Thickness Lead Spacer