Learn More
Specac™ Omni-Cell™ Fused Silica Liquid Cell Windows Rectangular
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
Description
Fused silica rectangular liquid cell windows for use in the Omni-Cell GS01800. 41mm x 23mm and 2mm thick.
Specifications
Specifications
| For Use With (Equipment) | Omni-Cell GS01800 |
| Description | 41 mm x 23 mm and 2 mm thick |