missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Omni-Cell™ CaF2 Sealed Window Unit 1.00mm
Click to view available options
Type:
Omni-Cell CaF2 Sealed Window Unit 1.00mm
Description
Permanently sealed Omni-Cell GS01800 with CaF2 rectangular windows and 1.00mm thickness lead spacer. Cannot be disassembled. Ideal for volatile liquids, quantitative applications and low viscosity liquids.
Specifications
Specifications
| Type | Omni-Cell CaF2 Sealed Window Unit 1.00mm |
| Description | 1.00 mm Thickness Lead Spacer |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction