missing translation for 'onlineSavingsMsg'
Learn More

Specac™ Omni-Cell™ CaF2 Mull Windows Circular 25mm Diameter

Product Code. 15269837
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
This item is not returnable. View return policy

Product Code. 15269837

Brand: Specac™ GS01832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
CaF2 circular mull cell windows for use in the Omni-Cell GS01800. 25 mm diameter and 4mm thick.

Specifications

For Use With (Equipment) Omni-Cell GS01800
Description 4 mm thick
Diameter (Metric) 25 mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.