Learn More
Specac™ Omni-Cell™ CaF2 Liquid Cell Windows Rectangular
Product Code. 15269827
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Omni-Cell GS01800
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15269827 Omni-Cell GS01800 Each
1 options
Product Code. 15269827 Supplier Specac™ Supplier No. GS01812
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
CaF2 rectangular Liquid Cell windows for use in the Omni-Cell GS01800. 41mm x 23mm and 4mm thick.
Specifications
For Use With (Equipment) Omni-Cell GS01800
Description 41 mm x 23 mm and 4 mm thick