missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Omni-Cell™ BaF2 Sealed Window Unit 0.20mm
Description
Permanently sealed Omni-Cell GS01800 with BaF2 rectangular windows and 0.20mm thickness lead spacer. Cannot be disassembled. Ideal for volatile liquids, quantitative applications and low viscosity liquids.
Specifications
Specifications
| Type | Omni-Cell BaF2 Sealed Window Unit 0.20mm |
| Description | 0.20 mm Thickness Lead Spacer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?