missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Omics Pouch

Product Code. 30057716 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Tissue Disaggregator
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
30057716 Tissue Disaggregator Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057716 Supplier Cytiva Supplier No. 29726921
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Single-use, multi-compartment pouch with tubing to allow the addition of digestive enzyme solutions.

  • Single-use
  • Multi-compartment
  • With tubing
TRUSTED_SUSTAINABILITY

Specifications

Product Type Pouch
Autoclavable Not Autoclavable
For Use With (Equipment) Tissue Disaggregator

For research use only (RUO). Not for diagnostic use.

Product Title
Select an issue

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.