missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Omics Clamp

Product Code. 30057714 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
Tissue Disaggregator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
30057714 Tissue Disaggregator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057714 Supplier Cytiva Supplier No. 29509355
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides a secure seal after sample insertion and supports the bag during tissue dissociation.

  • Provides a secure seal
  • Supports the bag during tissue dissociation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Clamp
Autoclavable Not Autoclavable
For Use With (Equipment) Tissue Disaggregator

For research use only (RUO). Not for diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.