missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole Sulfone, TRC

Product Code. 17913135
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913135 10 mg 10mg
17994205 25 mg 25mg
17984255 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913135 Supplier Toronto Research Chemicals Supplier No. TRCO63502510MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88546-55-8
Molecular Formula C17 H19 N3 O4 S
Molecular Weight (g/mol) 361.42
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulphonyl]-1H-benzimidazole, Esomeprazole Magnesium Trihydrate Imp. D (EP), Omeprazole Imp. D (EP), Omeprazole Sulphone, 1H-Benzimidazole, 6-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]- (ACI), 1H-Benzimidazole, 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]- (9CI), 6-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole (ACI), 2-[(3,5-Dimethyl-4-methoxy-2-pyridyl)methylsulfonyl]-5-methoxy-1H-benzimidazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole, H 168/66, Omeprazole sulfone
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfonyl]-1H-benzimidazole
SMILES COc1ccc2[nH]c(nc2c1)S(=O)(=O)Cc3ncc(C)c(OC)c3C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H19N3O4S/c1-10-8-18-15(11(2)16(10)24-4)9-25(21,22)17-19-13-6-5-12(23-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)
Formula Weight 361.1096
Percent Purity >95%
Chemical Name or Material Omeprazole sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.