missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole, MedChemExpress

Product Code. 16448106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448106 100 mg 100mg
16402026 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448106 Supplier Medchem Express Supplier No. HYB0113
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Omeprazole (H 16868), a proton pump inhibitor (PPI), is available for treatment of acid-related gastrointestinal disorders. Omeprazole shows competitive inhibition of CYP2C19 activity with a Ki of 2 to 6 μM. Omeprazole also inhibits growth of Gram-positive and Gram-negative bacteria.Omeprazole is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).

Omeprazole (H 16868), a proton pump inhibitor (PPI), is available for treatment of acid-related gastrointestinal disorders. Omeprazole shows competitive inhibition of CYP2C19 activity with a Ki of 2 to 6 μM. Omeprazole also inhibits growth of Gram-positive and Gram-negative bacteria.Omeprazole is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).

Specifications

Chemical Name or Material Omeprazole
CAS 73590-58-6
Color White
Purity Grade Notes Research
Molecular Formula C17H19N3O3S
Quantity 100 mg
Synonym H 16868
Solubility Information DMSO : ≥ 100 mg/mL (289.50 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(C1=NC2=CC=C(OC)C=C2N1)CC3=NC=C(C)C(OC)=C3C
Molecular Weight (g/mol) 345.42
Formula Weight 345.42
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.