missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole EP Impurity H, Mikromol™

Product Code. 30052306
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052306 25 mg 25mg
17903856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052306 Supplier Mikromol Supplier No. MM0095.150025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Omeprazole EP Impurity H
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(RS)-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole
CAS 863029-89-4
Analyte or Component Names 2-[(RS)-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H16 Cl N3 O2 S
Certifications/Compliance ISO 17025
Synonym 2-[(RS)-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1H-benzimidazole, Omeprazole Imp. H (EP), Omeprazole Impurity H
InChI Formula InChI=1S/C16H16ClN3O2S/c1-9-7-18-14(10(2)15(9)17)8-23(21)16-19-12-5-4-11(22-3)6-13(12)20-16/h4-7H,8H2,1-3H3,(H,19,20)
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3ncc(C)c(Cl)c3C
IUPAC Name 2-[(4-chloro-3,5-dimethylpyridin-2-yl)methylsulfinyl]-5-methoxy-1H-benzimidazole
Molecular Weight (g/mol) 349.84
Quantity 25 mg
Formula Weight 349.0652 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.