missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole Acid Disodium Salt, TRC

Product Code. 30427092
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427092 1 mg
30416299 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427092 Supplier Toronto Research Chemicals Supplier No. TRCO6350051MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120003-84-1
Molecular Formula C17H15N3Na2O5S
Molecular Weight (g/mol) 419.36
Synonym 4-Methoxy-6-[[(6-methoxy-1H-benzimidazol-2-yl)sulfinyl]methyl]-5-methyl-3-pyridinecarboxylic Acid Sodium Salt
IUPAC Name disodium;4-methoxy-6-[(5-methoxybenzimidazol-1-id-2-yl)sulfinylmethyl]-5-methylpyridine-3-carboxylate
SMILES [Na+].[Na+].COc1ccc2[n-]c(nc2c1)S(=O)Cc3ncc(C(=O)[O-])c(OC)c3C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H17N3O5S.2Na/c1-9-14(18-7-11(16(21)22)15(9)25-3)8-26(23)17-19-12-5-4-10(24-2)6-13(12)20-17;;/h4-7H,8H2,1-3H3,(H2,19,20,21,22);;/q;2*+1/p-2
Formula Weight 419.0528
Chemical Name or Material Omeprazole Acid Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.