missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OMD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598027
This item is not returnable.
View return policy
Description
OMD Polyclonal Antibody for Western Blot, IHC (P), ELISA
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.
Specifications
| OMD | |
| Polyclonal | |
| Unconjugated | |
| OMD | |
| Keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; Osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
| Rabbit | |
| Protein G | |
| RUO | |
| 4958 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.46 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q99983 | |
| OMD | |
| Recombinant Human Osteomodulin protein (339-411AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction