missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ombrabulin hydrochloride, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
Ombrabulin hydrochloride is a derivative of CA-4 phosphate, which is known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells.
Specifications
Specifications
| Chemical Name or Material | Ombrabulin hydrochloride |
| CAS | 253426-24-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H27ClN2O6 |
| Quantity | 100 mg |
| Synonym | AVE8062 hydrochlorideAC7700 hydrochloride |
| Solubility Information | H2O : 20 mg/mL (45.57 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CC(/C=C\C2=CC(OC)=C(OC)C(OC)=C2)=CC=C1OC)[C@@H](N)CO.[H]Cl |
| Molecular Weight (g/mol) | 438.9 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction