missing translation for 'onlineSavingsMsg'
Learn More

Omadacycline tosylate, MedChemExpress

Product Code. 16486218
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486218 10 mg 10mg
16472347 50 mg 50mg
16428817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486218 Supplier Medchem Express Supplier No. HY14865B
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Omadacycline (PTK 0796) tosylate, a first-in-class orally active aminomethylcycline antibacterial, is a member of the tetracycline class of antibiotics.

Omadacycline tosylate acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline tosylate possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline tosylate can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.

Specifications

Chemical Name or Material Omadacycline tosylate
CAS 1075240-43-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C36H48N4O10S
Quantity 10 mg
Synonym PTK 0796 tosylateAmadacycline tosylate
Solubility Information H2O : 12.5 mg/mL (17.15 mM; Need ultrasonic)
SMILES O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.OS(=O)(C5=CC=C(C)C=C5)=O
Molecular Weight (g/mol) 728.85
Formula Weight 728.85
Percent Purity 99.37%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.