missing translation for 'onlineSavingsMsg'
Learn More

Omadacycline hydrochloride, MedChemExpress

Product Code. 16496218
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496218 10 mg 10mg
16482347 50 mg 50mg
16438817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496218 Supplier Medchem Express Supplier No. HY14865C
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Omadacycline (PTK 0796) hydrochloride, a first-in-class orally active aminomethylcycline antibacterial, is a member of the tetracycline class of antibiotics.

Omadacycline hydrochloride acts through the inhibition of bacterial protein synthesis by binding to the 30S ribosomal subunit. Omadacycline hydrochloride possesses broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. Omadacycline hydrochloride can be used for the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.

Specifications

Chemical Name or Material Omadacycline hydrochloride
CAS 1196800-39-1
Color Dark Brown
Purity Grade Notes Research
Molecular Formula C29H40N4O7.HCl
Quantity 10 mg
Synonym PTK0796 hydrochloride Amadacycline hydrochloride
Solubility Information DMSO : 105 mg/mL (177.03 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (84.30 mM)
SMILES O=C(C1=C(O)[C@@H](N(C)C)[C@@](C[C@@]2([H])C(C(C3=C(O)C(CNCC(C)(C)C)=CC(N(C)C)=C3C2)=O)=C4O)([H])[C@@]4(O)C1=O)N.Cl
Molecular Weight (g/mol) 593.11
Formula Weight 593.11
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.