Learn More
Invitrogen™ OMA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113975
Description
Immunogen sequence: WAICPRDSLA LLCQWIQSKL QEYMFNRPYS RKLEAEADKI GLLLAAKACA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 85%; Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 86%; Rat: 86%; Yeast: 83%; Zebrafish: 85%.
OMA1 (Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family. It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants, and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.
Specifications
| OMA1 | |
| Polyclonal | |
| Unconjugated | |
| OMA1 | |
| 2010001O09Rik; DAB1; metalloendopeptidase OMA1, mitochondrial; metalloprotease related protein 1; metalloprotease-related protein 1; MPRP1; MPRP-1; Oma1; OMA1 homolog, zinc metallopeptidase; OMA1 homolog, zinc metallopeptidase (S. cerevisiae); OMA1 zinc metallopeptidase; OMA1 zinc metallopeptidase homolog; overlapping activity with M-AAA protease; overlapping with the m-AAA protease 1 homolog; peptidase; RGD1304821; YKR087C; zinc metallopeptidase homolog; zinc metallopeptidase OMA1; ZMPOMA1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 115209 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96E52 | |
| OMA1 | |
| Synthetic peptide directed towards the middle region of human OMA1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.