Learn More
OMA1 Mouse anti-Human, Unconjugated, Clone: 1H4D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67449-1-IG-150UL
This item is not returnable.
View return policy
Description
The OMA1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human OMA1. This antibody recognizes human, mouse antigen. The OMA1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| OMA1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, ELISA | |
| 1H4D12 | |
| Unconjugated | |
| 150 μL | |
| 115209 | |
| IgG1 |