Learn More
Thermo Scientific™ Olympus™ 60X Oil Objective, X-Apo, 1.42NA/0.15WD
Shop All Thermo Scientific ProductsDescription
This 60X apochromat objective is from the new Olympus X line with exceptional optical performance. Through new manufacturing technology, the X-Apo objectives offer improved performance compared to standard Apo objectives in several critical areas: larger numerical aperture (NA), better image flatness, and a wider range of chromatic correction of 400–1,000 nm. These objectives deliver superb brightness and resolution across a large field of view for applications where the best possible image quality is desired. The Olympus objectives are compatible with all EVOS cell imaging systems. Our wide selection of EVOS and Olympus objectives satisfies needs across the spectrum of magnifications and optical specifications.
Additional characteristics of this Olympus objective:
- Magnification: 60X
- Numerical Aperture: 1.42
- Working Distance: 0.15 mm
- Immersion: oil
- Superb image quality
Image quality
Microscope objectives may be the most important components of a microscope because they are responsible for primary image formation. High image quality is crucial to experimental success and often a requirement for publication; Olympus objectives afford that quality across the visible spectrum to near infrared light. This performance results from years of lens manufacture experience, and the broad selection of objectives means that you can choose the optimal objective for your imaging requirements.
Objective classes
Achromat objectives are perfect for general applications, with standard correction of color and focus.
Fluorite objectives deliver excellent resolution and are made with higher numerical apertures than achromat objectives, resulting in brighter fluorescence signal and higher contrast imaging. The higher optical quality greatly reduces optical aberrations, and corrections for color and focus are at higher levels than achromat objectives. Fluorite objectives are ideally suited for fluorescence and demanding transmitted light applications, where the higher contrast make them ideally suited for color imaging.
Apochromat objectives are manufactured to the highest levels of resolution, fluorescence brightness, and contrast; chromatic aberrations are almost eliminated. They are recommended for the most demanding applications, particularly at magnifications of 60x and above. Apochromatic objectives are the best choice for the capture of color images in white light.
Brightfield contrast versus phase contrast objectives
Brightfield is the most basic form of light microscopy and is accomplished by sample absorption of light. A higher density area in a sample will absorb more light, thus increasing contrast in those areas. Phase contrast objectives are most useful for hard to see, translucent specimens. This method of contrast is accomplished by converting phase shifts, caused by light passing through a translucent specimen, into brightness changes (i.e., contrast).
Long working distance versus coverslip-corrected objectives
Long working distance (LWD) objectives are optimized for use through vessels with a nominal wall thickness of 0.9-1.5 mm. This includes vessels commonly used in cell culture and cell-based assays, such as slides, cell culture dishes and flasks, microtiter well plates, etc. Coverslip-corrected objectives are optimized for use through #1.5 coverslips (thickness approximately 0.17 mm). These objectives have a higher magnification/NA ratio and provide higher resolution compared to LWD objectives.
Specifications
Specifications
| Type | Objective Lens |
| Product Line | Olympus |
| For Use With (Equipment) | EVOS M5000, EVOS M7000 and Other Imaging Systems |
| Includes | Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates |
| Quantity | Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
