missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLT-2 Antibody (2564C), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10059CL1
This item is not returnable.
View return policy
Description
OLT-2 Monoclonal antibody specifically detects OLT-2 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| OLT-2 | |
| Monoclonal | |
| CoraFluor 1 | |
| OSCAR-Like Transcript-2 Protein, T Cell-Interacting, Activating Receptor On Myeloid Cells, T Cell-Interacting, Activating Receptor On Myeloid Cells 1, TARM1, T-Cell-Interacting, Activating Receptor On Myeloid Cells Protein 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human OLT-2/TARM-1, Met1-Val235, Accession # B6A8C7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 2564C | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 441864 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction