missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | OLR1 |
|---|---|
| Concentration | 0.46 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16839173
|
Proteintech
11837-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16829173
|
Proteintech
11837-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a low density lipoprotein receptor that belongs to the C-type lectin superfamily. This gene is regulated through the cyclic AMP signaling pathway. The encoded protein binds, internalizes and degrades oxidized low-density lipoprotein. This protein may be involved in the regulation of Fas-induced apoptosis. This protein may play a role as a scavenger receptor. Mutations of this gene have been associated with atherosclerosis, risk of myocardial infarction, and may modify the risk of Alzheimer′s disease. Alternate splicing results in multiple transcript variants.Specifications
| OLR1 | |
| Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P78380 | |
| 4973 | |
| OLR1 Fusion Protein Ag2437 | |
| Primary | |
| -20°C | |
| OLR1 |
| 0.46 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CLEC8A, hLOX 1, Lectin like oxLDL receptor 1, LOX 1, LOX1, OLR1, Ox LDL receptor 1, SCARE1 | |
| OLR1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title