missing translation for 'onlineSavingsMsg'
Learn More

Olprinone Hydrochloride, MedChemExpress

Product Code. 16442147
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442147 50 mg 50mg
16416008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442147 Supplier Medchem Express Supplier No. HY14254
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Olprinone (Loprinone) Hydrochloride is a potent phosphodiesterase (PDE) 3 inhibitor, with IC50s of 150, 100, 0.35 and 14 μM for PDE1, PDE2, PDE3 and PDE4, respectively. Olprinone Hydrochloride is used for the research of heart failure due to its positive inotropic and vasodilative effects. Anti-inflammatory activity.

Olprinone (Loprinone) Hydrochloride is a potent phosphodiesterase (PDE) 3 inhibitor, with IC50s of 150, 100, 0.35 and 14 μM for PDE1, PDE2, PDE3 and PDE4, respectively. Olprinone Hydrochloride is used for the research of heart failure due to its positive inotropic and vasodilative effects. Anti-inflammatory activity.

Specifications

Chemical Name or Material Olprinone Hydrochloride
CAS 119615-63-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H11ClN4O
Quantity 50 mg
Synonym Loprinone Hydrochloride
Solubility Information H2O : ≥ 7.69 mg/mL (26.82 mM)
SMILES CC(N1)=C(C=C(C#N)C1=O)C2=CN3C(C=C2)=NC=C3.[H]Cl
Molecular Weight (g/mol) 286.72
Formula Weight 286.72
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.