missing translation for 'onlineSavingsMsg'
Learn More

Olopatadine, TRC

Product Code. 30412277
Change view
Click to view available options
Quantity:
25 mg
100 mg
Product Code. Quantity
30412277 100 mg
30418382 25 mg
2 options
This item is not returnable. View return policy

Product Code. 30412277

Brand: Toronto Research Chemicals TRCO574995100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113806-05-6
Formula Weight 337.1678
InChI Formula InChI=1S/C21H23NO3/c1-22(2)11-5-8-18-17-7-4-3-6-16(17)14-25-20-10-9-15(12-19(18)20)13-21(23)24/h3-4,6-10,12H,5,11,13-14H2,1-2H3,(H,23,24)/b18-8-
IUPAC Name 2-[(11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetic acid
Molecular Formula C21H23NO3
Molecular Weight (g/mol) 337.41
SMILES CN(C)CC\C=C/1\c2ccccc2COc3ccc(CC(=O)O)cc13
Synonym (Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic Acid, 11-[(Z)-3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic Acid, Opatanol, Pataday, Patanol
Chemical Name or Material Olopatadine

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.