missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olopatadine Ethyl Ester, Mikromol™
Description
Olopatadine Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Olopatadine Ethyl Ester |
| CAS | 113806-03-4 |
| Analyte or Component Names | Olopatadine Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Olopatadine Ethyl Ester, (11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid ethyl ester, Dibenz[b,e]oxepin-2-acetic acid, 11-[3-(dimethylamino)propylidene]-6,11-dihydro-, ethyl ester, (Z)- |
| InChI Formula | InChI=1S/C23H27NO3/c1-4-26-23(25)15-17-11-12-22-21(14-17)20(10-7-13-24(2)3)19-9-6-5-8-18(19)16-27-22/h5-6,8-12,14H,4,7,13,15-16H2,1-3H3/b20-10- |
| SMILES | CCOC(=O)Cc1ccc2OCc3ccccc3\C(=C\CCN(C)C)\c2c1 |
| IUPAC Name | ethyl 2-[(11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?