missing translation for 'onlineSavingsMsg'
Learn More

Olodaterol Hydrochloride, TRC

Product Code. 30009461
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009461 1 mg 1mg
30009181 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009461 Supplier Toronto Research Chemicals Supplier No. TRCO2620001MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869477-96-3
Molecular Formula C21H27ClN2O5
Molecular Weight (g/mol) 422.9
Synonym 2H-1,4-Benzoxazin-3(4H)-one, 6-hydroxy-8-[(1R)-1-hydroxy-2-[[2-(4-methoxyphenyl)-1,1-dimethylethyl]amino]ethyl]-, hydrochloride (1:1), 2H-1,4-Benzoxazin-3(4H)-one, 6-hydroxy-8-[(1R)-1-hydroxy-2-[[2-(4-methoxyphenyl)-1,1-dimethylethyl]amino]ethyl]-, monohydrochloride (9CI), BI 1744 hydrochloride, Olodaterol hydrochloride
SMILES Cl.COc1ccc(CC(C)(C)NC[C@H](O)c2cc(O)cc3NC(=O)COc23)cc1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H26N2O5.ClH/c1-21(2,10-13-4-6-15(27-3)7-5-13)22-11-18(25)16-8-14(24)9-17-20(16)28-12-19(26)23-17;/h4-9,18,22,24-25H,10-12H2,1-3H3,(H,23,26);1H/t18-;/m0./s1
Formula Weight 422.1608
Percent Purity >95%
Chemical Name or Material Olodaterol Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.