missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olodanrigan, MedChemExpress
Description
Olodanrigan (EMA401) is a highly selective, orally active, peripherally restricted angiotensin II type 2 receptor (AT2R) antagonist. It is under development as a neuropathic pain therapeutic agent. Olodanrigan (EMA401) analgesic action appears to involve inhibition of augmented AngII/AT2R induced p38 and p42/p44 MAPK activation, and hence inhibition of DRG neuron hyperexcitability and sprouting of DRG neurons.
Specifications
Specifications
| Chemical Name or Material | Olodanrigan |
| CAS | 1316755-16-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H29NO5 |
| Quantity | 5 mg |
| Synonym | EMA401 PD-126055 |
| Solubility Information | DMSO : ≥ 34 mg/mL (66.98 mM) ∣H2O : 0.1 mg/mL (0.20 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1N(C(C(C2=CC=CC=C2)C3=CC=CC=C3)=O)CC4=C(C(OCC5=CC=CC=C5)=C(OC)C=C4)C1)O |
| Molecular Weight (g/mol) | 507.58 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?