missing translation for 'onlineSavingsMsg'
Learn More

Olmesartan, Tocris Bioscience™

Product Code. 18025002
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18025002

Brand: Tocris Bioscience 4616/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent AT1 antagonist; metabolite of Olmesartan medoxomil (Cat. No. 4620)

Potent angiotensin II type I (AT1) receptor antagonist (IC50 = 6.7 nM at hAT1 receptors). Improves cognitive function in a mouse model of hypertension- and hyperlipidemia-associated cognitive decline; ameliorates arterial stiffness in a rat model of chronic renal failure.
TRUSTED_SUSTAINABILITY

Specifications

CAS 144689-24-7
Molecular Formula C24H26N6O3
MDL Number MFCD00914967
Quantity 50 mg
Synonym olmesartan, olmesartan acid, votum, olme sartan, unii-8w1iqp3u10, chembl1516, 4-2-hydroxypropan-2-yl-2-propyl-1-4-2-1h-1,2,3,4-tetrazol-5-yl phenyl phenyl methyl-1h-imidazole-5-carboxylic acid, 1-2'-1h-tetrazol-5-yl-1,1'-biphenyl-4-yl methyl-4-2-hydroxypropan-2-yl-2-propyl-1h-imidazole-5-carboxylic acid, 1h-imidazole-5-carboxylic acid, 4-1-hydroxy-1-methylethyl-2-propyl-1-2'-1h-tetrazol-5-yl 1,1'-biphenyl-4-yl methyl, 4-1-hydroxy-1-methylethyl-2-propyl-1-2'-2h-tetrazol-5-yl 1,1'-biphenyl-4-yl methyl-1h-imidazole-5-carboxylic acid
InChI Key VTRAEEWXHOVJFV-UHFFFAOYSA-N
SMILES CCCC1=NC(=C(N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)C(O)=O)C(C)(C)O
IUPAC Name 4-(2-hydroxypropan-2-yl)-2-propyl-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-imidazole-5-carboxylic acid
Molecular Weight (g/mol) 446.51
PubChem CID 158781
ChEBI CHEBI:48416
Formula Weight 446.5
Percent Purity >99%
Chemical Name or Material Olmesartan
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.